Assay Details
Gene Symbol: Dchs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232535.2 | XP_006232597.1 | 12 - 13 | 6013 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
