Assay Details
Gene Symbol: Dchs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232535.2 | XP_006232597.1 | 15 - 16 | 7201 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
