Assay Details
Gene Symbol: Pbxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100976.1 | NP_001094446.1 | 6 - 7 | 692 | 148 | |
XM_006232668.1 | XP_006232730.1 | 7 - 8 | 819 | 148 | ||
XM_006232669.1 | XP_006232731.1 | 7 - 8 | 798 | 148 | ||
XM_008761174.1 | XP_008759396.1 | 7 - 8 | 843 | 148 | ||
XM_008761175.1 | XP_008759397.1 | 7 - 8 | 843 | 148 | ||
GenBank mRNA | BC129100.1 | - | 6 - 7 | 692 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
