Assay Details
Gene Symbol: Pbxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100976.1 | NP_001094446.1 | 8 - 9 | 893 | 68 | |
XM_006232668.1 | XP_006232730.1 | 9 - 10 | 1020 | 68 | ||
XM_006232669.1 | XP_006232731.1 | 9 - 10 | 999 | 68 | ||
XM_008761174.1 | XP_008759396.1 | 9 - 10 | 1044 | 68 | ||
GenBank mRNA | BC129100.1 | - | 8 - 9 | 893 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
