Assay Details
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057140.1 | NP_476481.1 | 11 - 12 | 1364 | 120 | |
GenBank mRNA | BC083859.1 | - | 11 - 12 | 1364 | 120 | |
D14418.1 | - | 2 - 3 | 122 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
