Assay Details
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057140.1 | NP_476481.1 | 14 - 15 | 1755 | 158 | |
GenBank mRNA | BC083859.1 | - | 14 - 15 | 1755 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P427523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139104 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P427523 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P427523 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P427523 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P297119 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P427523 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700511794 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32694 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31286 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34127 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700511794 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10703488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703494 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703489 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703483 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703486 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703492 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703482 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703480 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703484 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703493 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703485 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703487 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703481 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703490 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703491 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10703479 | - | - |
Rat Genome 230 2.0 Array | 1375245_at | - | - |
Rat Genome 230 2.0 Array | 1375245_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381024_at | - | - |
Rat Genome 230 2.0 Array | 1381024_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375245_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375245_at | - | - |
Affymetrix HT Rat Focus Array | 1381024_at | - | - |
Affymetrix HT Rat Focus Array | 1381024_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100004116.rn.1 | TC0100000643.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100004117.rn.1 | TC0100000643.rn.1 | RTA TC |
Rat Expression Set 230 | 1375245_PM_at | - | - |
Rat Expression Set 230 | 1375245_at | - | - |
Rat Expression Set 230 | 1381024_at | - | - |
Rat Expression Set 230 | 1381024_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10703479 | - | - |
Rat Genome U34 Set | D14418_at | - | - |
Rat Genome U34 Set | rc_AA800669_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371662 | - | - |
RatRef-12 Expression BeadChip | ILMN_63422 | - | - |
