Assay Details
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057140.1 | NP_476481.1 | 1 - 2 | 74 | 72 | |
GenBank mRNA | BC083859.1 | - | 1 - 2 | 74 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
