Assay Details
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057140.1 | NP_476481.1 | 8 - 9 | 993 | 127 | |
GenBank mRNA | BC083859.1 | - | 8 - 9 | 993 | 7126254 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
