Assay Details
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004198.1 | NP_001004198.1 | 2 - 3 | 318 | 109 | |
XM_006227980.2 | XP_006228042.1 | 2 - 3 | 261 | 109 | ||
XM_008758747.1 | XP_008756969.1 | 2 - 3 | 261 | 109 | ||
XM_008758748.1 | XP_008756970.1 | 2 - 3 | 469 | 109 | ||
GenBank mRNA | BC081939.1 | - | 2 - 3 | 318 | 7115389 | 109 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC081939.1,NM_001004198.1,XM_006227980.2,XM_008758747.1,XM_008758748.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
