Assay Details
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004198.1 | NP_001004198.1 | 3 - 4 | 702 | 74 | |
XM_006227980.2 | XP_006228042.1 | 3 - 4 | 645 | 74 | ||
XM_008758747.1 | XP_008756969.1 | 3 - 4 | 645 | 74 | ||
XM_008758748.1 | XP_008756970.1 | 3 - 4 | 853 | 74 | ||
GenBank mRNA | BC081939.1 | - | 3 - 4 | 702 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
