Assay Details
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004198.1 | NP_001004198.1 | 6 - 7 | 1041 | 153 | |
XM_006227980.2 | XP_006228042.1 | 6 - 7 | 984 | 153 | ||
GenBank mRNA | BC081939.1 | - | 6 - 7 | 1041 | 7115389 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
