Assay Details
Gene Symbol: Akr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138884.1 | NP_620239.1 | 3 - 4 | 415 | 119 | |
XM_006236285.1 | XP_006236347.1 | 3 - 4 | 435 | 119 | ||
GenBank mRNA | D17309.1 | - | 3 - 4 | 415 | 119 | |
FQ211413.1 | - | 3 - 4 | 433 | 119 | ||
FQ219185.1 | - | 3 - 4 | 434 | 119 | ||
FQ219421.1 | - | 3 - 4 | 378 | 119 | ||
FQ219730.1 | - | 3 - 4 | 378 | 119 | ||
S80431.1 | - | 3 - 4 | 387 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
