Assay Details
Gene Symbol: Ubn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134553.1 | NP_001128025.1 | 17 - 18 | 4191 | 67 | |
XM_006236306.2 | XP_006236368.1 | 17 - 18 | 5280 | 67 | ||
XM_006236307.2 | XP_006236369.1 | 16 - 17 | 5229 | 67 | ||
XM_006236308.2 | XP_006236370.1 | 16 - 17 | 5223 | 67 | ||
GenBank mRNA | FQ212455.1 | - | 3 - 4 | 3763 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P009415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2960 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P009415 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P009415 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P009415 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P177162 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10854603 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854613 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854609 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854615 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854607 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854605 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854608 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854612 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854618 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854604 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854602 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854611 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854617 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854616 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854610 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854619 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854606 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854614 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10854601 | - | - |
Rat Genome 230 2.0 Array | 1373371_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1397959_at | - | - |
Rat Genome 230 2.0 Array | 1397959_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373371_a_at | - | - |
Affymetrix HT Rat Focus Array | 1397959_at | - | - |
Affymetrix HT Rat Focus Array | 1397959_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400004794.rn.1 | TC0400000662.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400004793.rn.1 | TC0400000662.rn.1 | RTA TC |
Rat Expression Set 230 | 1397959_PM_at | - | - |
Rat Expression Set 230 | 1397959_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10854601 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21428038 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357031 | - | - |
