Assay Details
Gene Symbol: Ubn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134553.1 | NP_001128025.1 | 4 - 5 | 1001 | 108 | |
XM_006236306.2 | XP_006236368.1 | 4 - 5 | 2084 | 108 | ||
XM_006236307.2 | XP_006236369.1 | 4 - 5 | 2084 | 108 | ||
XM_006236308.2 | XP_006236370.1 | 4 - 5 | 2084 | 108 | ||
XM_006236311.2 | XP_006236373.1 | 4 - 5 | 2084 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
