Assay Details
Gene Symbol: Gstk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181371.2 | NP_852036.1 | 1 - 2 | 108 | 94 | |
XM_006236391.2 | XP_006236453.1 | 1 - 2 | 179 | 94 | ||
GenBank mRNA | FJ179400.1 | - | 1 - 2 | 74 | 94 | |
FQ211030.1 | - | 1 - 2 | 126 | 94 | ||
FQ214179.1 | - | 1 - 2 | 191 | 94 | ||
S83436.1 | - | 1 - 2 | 108 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
