Assay Details
Gene Symbol: Impdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108619.1 | NP_001102089.1 | 12 - 13 | 1431 | 57 | |
XM_006236208.1 | XP_006236270.1 | 12 - 13 | 1653 | 57 | ||
XM_006236210.1 | XP_006236272.1 | 12 - 13 | 1653 | 57 | ||
XM_008762791.1 | XP_008761013.1 | 12 - 13 | 1656 | 57 | ||
XM_008762792.1 | XP_008761014.1 | 11 - 12 | 1581 | 57 | ||
XM_008762793.1 | XP_008761015.1 | 12 - 13 | 1656 | 57 | ||
XM_008762794.1 | XP_008761016.1 | 11 - 12 | 1581 | 57 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
