Assay Details
Gene Symbol: Impdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108619.1 | NP_001102089.1 | 5 - 6 | 521 | 65 | |
XM_006236208.1 | XP_006236270.1 | 5 - 6 | 674 | 65 | ||
XM_006236210.1 | XP_006236272.1 | 5 - 6 | 674 | 65 | ||
XM_008762791.1 | XP_008761013.1 | 5 - 6 | 677 | 65 | ||
XM_008762793.1 | XP_008761015.1 | 5 - 6 | 677 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
