Assay Details
Gene Symbol: Tspan12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015026.1 | NP_001015026.1 | 5 - 6 | 756 | 91 | |
XM_008762784.1 | XP_008761006.1 | 6 - 7 | 678 | 91 | ||
XM_008762785.1 | XP_008761007.1 | 6 - 7 | 607 | 91 | ||
XM_008762786.1 | XP_008761008.1 | 6 - 7 | 606 | 91 | ||
XM_008762787.1 | XP_008761009.1 | 6 - 7 | 607 | 91 | ||
XM_008762788.1 | XP_008761010.1 | 5 - 6 | 626 | 91 | ||
XM_008762789.1 | XP_008761011.1 | 5 - 6 | 889 | 91 | ||
XM_008762790.1 | XP_008761012.1 | 5 - 6 | 889 | 91 | ||
GenBank mRNA | BC092623.1 | - | 5 - 6 | 756 | 7313631 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
