Assay Details
Gene Symbol: Tspan12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015026.1 | NP_001015026.1 | 7 - 8 | 1002 | 140 | |
XM_008762784.1 | XP_008761006.1 | 8 - 9 | 924 | 140 | ||
XM_008762785.1 | XP_008761007.1 | 8 - 9 | 853 | 140 | ||
XM_008762786.1 | XP_008761008.1 | 8 - 9 | 852 | 140 | ||
XM_008762787.1 | XP_008761009.1 | 8 - 9 | 853 | 140 | ||
XM_008762788.1 | XP_008761010.1 | 7 - 8 | 872 | 140 | ||
GenBank mRNA | BC092623.1 | - | 7 - 8 | 1002 | 7313631 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P329090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152163 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P329090 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P329090 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P329090 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P665848 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P329090 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P473459 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40323 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59907 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94899 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35308 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59848 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73294 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10861223 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861216 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861215 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861217 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861222 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861221 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861220 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861219 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861214 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861218 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10861213 | - | - |
Rat Genome 230 2.0 Array | 1372455_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372455_at | - | - |
Affymetrix HT Rat Focus Array | 1372455_at | - | - |
Affymetrix HT Rat Focus Array | 1372455_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400017828.rn.1 | TC0400002523.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400017827.rn.1 | TC0400002523.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400017830.rn.1 | TC0400002523.rn.1 | RTA TC |
Rat Expression Set 230 | 1372455_at | - | - |
Rat Expression Set 230 | 1372455_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10861213 | - | - |
Rat Genome U34 Set | rc_AI030542_at | - | - |
Rat Genome U34 Set | rc_AI145007_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20879231 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374598 | - | - |
