Assay Details
Gene Symbol: Tspan12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015026.1 | NP_001015026.1 | 1 - 2 | 327 | 91 | |
XM_008762784.1 | XP_008761006.1 | 2 - 3 | 249 | 91 | ||
XM_008762785.1 | XP_008761007.1 | 2 - 3 | 178 | 91 | ||
XM_008762786.1 | XP_008761008.1 | 2 - 3 | 177 | 91 | ||
XM_008762787.1 | XP_008761009.1 | 2 - 3 | 178 | 91 | ||
XM_008762789.1 | XP_008761011.1 | 1 - 2 | 460 | 91 | ||
XM_008762790.1 | XP_008761012.1 | 1 - 2 | 460 | 91 | ||
GenBank mRNA | BC092623.1 | - | 1 - 2 | 327 | 7313631 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC092623.1, NM_001015026.1, XM_008762784.1, XM_008762785.1, XM_008762786.1, XM_008762787.1, XM_008762789.1, XM_008762790.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
