Assay Details
Gene Symbol: Ssbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762822.1 | XP_008761044.1 | 2 - 3 | 267 | 80 | |
XM_008762823.1 | XP_008761045.1 | 2 - 3 | 203 | 80 | ||
XM_008762824.1 | XP_008761046.1 | 2 - 3 | 147 | 80 | ||
XM_008762825.1 | XP_008761047.1 | 2 - 3 | 135 | 80 | ||
XM_008762826.1 | XP_008761048.1 | 2 - 3 | 86 | 80 | ||
XM_008762827.1 | XP_008761049.1 | 2 - 3 | 267 | 80 | ||
XM_008762828.1 | XP_008761050.1 | 2 - 3 | 142 | 80 | ||
XM_008762829.1 | XP_008761051.1 | 1 - 2 | 66 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008762822.1, XM_008762823.1, XM_008762824.1, XM_008762825.1, XM_008762826.1, XM_008762827.1, XM_008762828.1, XM_008762829.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
