Assay Details
Gene Symbol: Ssbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183328.2 | NP_899157.1 | 4 - 5 | 313 | 70 | |
XM_008762822.1 | XP_008761044.1 | 5 - 6 | 559 | 70 | ||
XM_008762823.1 | XP_008761045.1 | 5 - 6 | 495 | 70 | ||
XM_008762824.1 | XP_008761046.1 | 5 - 6 | 439 | 70 | ||
XM_008762825.1 | XP_008761047.1 | 5 - 6 | 427 | 70 | ||
XM_008762826.1 | XP_008761048.1 | 5 - 6 | 378 | 70 | ||
XM_008762827.1 | XP_008761049.1 | 5 - 6 | 559 | 70 | ||
XM_008762828.1 | XP_008761050.1 | 5 - 6 | 434 | 70 | ||
XM_008762829.1 | XP_008761051.1 | 4 - 5 | 358 | 70 | ||
GenBank mRNA | M94557.1 | - | 4 - 5 | 367 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
