Assay Details
Gene Symbol: Ssbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762822.1 | XP_008761044.1 | 1 - 2 | 209 | 122 | |
XM_008762827.1 | XP_008761049.1 | 1 - 2 | 209 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008762822.1, XM_008762827.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
