Assay Details
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022535.2 | NP_071980.2 | 3 - 4 | 461 | 85 | |
GenBank mRNA | CK476875.1 | - | 3 - 4 | 482 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
