Assay Details
Gene Symbol: Ube2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172127.1 | NP_001165598.1 | 3 - 4 | 253 | 182 | |
GenBank mRNA | CB739657.1 | - | 1 - 2 | 69 | 182 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
