Assay Details
Gene Symbol: Ube2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172127.1 | NP_001165598.1 | 6 - 7 | 467 | 63 | |
GenBank mRNA | CB739657.1 | - | 4 - 5 | 283 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
