Assay Details
Gene Symbol: Tcaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749739.3 | XP_003749787.1 | 2 - 3 | 787 | 104 | |
XM_008762928.1 | XP_008761150.1 | 2 - 3 | 1116 | 104 | ||
XM_342675.6 | XP_342676.4 | 2 - 3 | 1116 | 104 | ||
GenBank mRNA | FQ211472.1 | - | 2 - 3 | 754 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
