Assay Details
Gene Symbol: Tcaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749739.3 | XP_003749787.1 | 3 - 4 | 1782 | 75 | |
XM_008762928.1 | XP_008761150.1 | 3 - 4 | 2111 | 75 | ||
XM_342675.6 | XP_342676.4 | 3 - 4 | 2111 | 75 | ||
GenBank mRNA | FQ211472.1 | - | 3 - 4 | 1749 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
