Assay Details
Gene Symbol: Tcaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749739.3 | XP_003749787.1 | 8 - 9 | 2970 | 85 | |
XM_342675.6 | XP_342676.4 | 8 - 9 | 3299 | 85 | ||
GenBank mRNA | FQ211472.1 | - | 8 - 9 | 2937 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
