Assay Details
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100708.1 | NP_001094178.1 | 6 - 7 | 855 | 80 | |
XM_006236198.2 | XP_006236260.1 | 6 - 7 | 903 | 80 | ||
XM_006236200.2 | XP_006236262.1 | 6 - 7 | 903 | 80 | ||
XM_008762772.1 | XP_008760994.1 | 3 - 4 | 961 | 80 | ||
XM_008762773.1 | XP_008760995.1 | 3 - 4 | 960 | 80 | ||
XM_008762774.1 | XP_008760996.1 | 6 - 7 | 901 | 80 | ||
GenBank mRNA | BC161956.1 | - | 6 - 7 | 855 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
