Assay Details
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100708.1 | NP_001094178.1 | 7 - 8 | 1010 | 90 | |
XM_006236198.2 | XP_006236260.1 | 7 - 8 | 1058 | 90 | ||
XM_006236200.2 | XP_006236262.1 | 7 - 8 | 1058 | 90 | ||
XM_008762772.1 | XP_008760994.1 | 4 - 5 | 1116 | 90 | ||
XM_008762773.1 | XP_008760995.1 | 4 - 5 | 1115 | 90 | ||
XM_008762774.1 | XP_008760996.1 | 7 - 8 | 1056 | 90 | ||
GenBank mRNA | BC161956.1 | - | 7 - 8 | 1010 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
