Assay Details
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012687.1 | NP_036819.1 | 7 - 8 | 822 | 58 | |
XM_008762759.1 | XP_008760981.1 | 7 - 8 | 883 | 58 | ||
XM_008762760.1 | XP_008760982.1 | 5 - 6 | 2280 | 58 | ||
XM_008762761.1 | XP_008760983.1 | 7 - 8 | 873 | 58 | ||
GenBank mRNA | D28773.1 | - | 7 - 8 | 822 | 58 | |
D31798.1 | - | 7 - 8 | 744 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
