Assay Details
Gene Symbol: Tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052800.1 | NP_434687.1 | 3 - 4 | 274 | 84 | |
GenBank mRNA | AW919734.1 | - | 1 - 2 | 111 | 84 | |
CV072953.1 | - | 3 - 4 | 273 | 84 | ||
FQ229684.1 | - | 3 - 4 | 314 | 84 | ||
FQ234584.1 | - | 284 | 84 | |||
FQ234769.1 | - | 284 | 84 | |||
M22642.1 | - | 3 - 4 | 271 | 84 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
