Assay Details
Gene Symbol: Tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052800.1 | NP_434687.1 | 5 - 6 | 458 | 88 | |
GenBank mRNA | AW919734.1 | - | 3 - 4 | 295 | 88 | |
CV072953.1 | - | 5 - 6 | 457 | 88 | ||
FQ234584.1 | - | 468 | 88 | |||
FQ234769.1 | - | 468 | 88 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
