Assay Details
Gene Symbol: Spata22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191824.1 | NP_001178753.1 | 3 - 4 | 237 | 117 | |
XM_006246762.2 | XP_006246824.1 | 4 - 5 | 443 | 117 | ||
XM_006246763.2 | XP_006246825.1 | 4 - 5 | 337 | 117 | ||
GenBank mRNA | AB236891.1 | - | 4 - 5 | 431 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
