Assay Details
Gene Symbol: Spata22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191824.1 | NP_001178753.1 | 7 - 8 | 895 | 116 | |
XM_006246762.2 | XP_006246824.1 | 8 - 9 | 1101 | 116 | ||
XM_006246763.2 | XP_006246825.1 | 8 - 9 | 995 | 116 | ||
GenBank mRNA | AB236891.1 | - | 8 - 9 | 1089 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
