Assay Details
Gene Symbol: Gdpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044238.1 | NP_001037703.1 | 3 - 4 | 425 | 75 | |
GenBank mRNA | BC105620.1 | - | 3 - 4 | 425 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
