Assay Details
Gene Symbol: Crlf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168612.1 | NP_001162083.1 | 1 - 2 | 357 | 142 | |
GenBank mRNA | AF072835.1 | - | 265 | 142 | ||
CB611803.1 | - | 357 | 143 | |||
CO555844.1 | - | 1 - 2 | 356 | 142 | ||
DY321186.1 | - | 1 - 2 | 350 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
