Assay Details
Gene Symbol: Crlf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168612.1 | NP_001162083.1 | 2 - 3 | 443 | 110 | |
GenBank mRNA | AF072835.1 | - | 351 | 110 | ||
CO555844.1 | - | 2 - 3 | 442 | 110 | ||
DY321186.1 | - | 2 - 3 | 436 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
