Assay Details
Gene Symbol: Crlf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168612.1 | NP_001162083.1 | 979 | 129 | ||
GenBank mRNA | AF072835.1 | - | 887 | 129 | ||
CV075881.1 | - | 229 | 129 | |||
DY560460.1 | - | 78 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
