Assay Details
Gene Symbol: Rpl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191589.1 | NP_001178518.1 | 5 - 6 | 723 | 133 | |
GenBank mRNA | FQ214717.1 | - | 5 - 6 | 742 | 133 | |
FQ215051.1 | - | 5 - 6 | 705 | 133 | ||
FQ216207.1 | - | 5 - 6 | 705 | 133 | ||
FQ216222.1 | - | 5 - 6 | 705 | 133 | ||
FQ216295.1 | - | 5 - 6 | 717 | 133 | ||
FQ216388.1 | - | 5 - 6 | 133 | |||
FQ216645.1 | - | 5 - 6 | 704 | 133 | ||
FQ216791.1 | - | 5 - 6 | 742 | 133 | ||
FQ224678.1 | - | 5 - 6 | 752 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
