Assay Details
Gene Symbol: Rpain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033060.1 | NP_001028232.1 | 1 - 2 | 84 | 118 | |
XM_006246668.2 | XP_006246730.1 | 1 - 2 | 448 | 118 | ||
GenBank mRNA | AY775322.1 | - | 1 - 2 | 84 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
