Assay Details
Gene Symbol: Rpain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033060.1 | NP_001028232.1 | 4 - 5 | 419 | 79 | |
XM_006246668.2 | XP_006246730.1 | 4 - 5 | 783 | 79 | ||
GenBank mRNA | AY775322.1 | - | 4 - 5 | 419 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
