Assay Details
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012140.1 | NP_001012140.1 | 2 - 3 | 315 | 97 | |
GenBank mRNA | BC086400.1 | - | 2 - 3 | 315 | 97 | |
FQ212591.1 | - | 3 - 4 | 365 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
