Assay Details
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012140.1 | NP_001012140.1 | 3 - 4 | 384 | 126 | |
GenBank mRNA | BC086400.1 | - | 3 - 4 | 384 | 126 | |
FQ212591.1 | - | 4 - 5 | 434 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
