Assay Details
Gene Symbol: Spns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109035.1 | NP_001102505.1 | 8 - 9 | 1213 | 100 | |
XM_006246786.2 | XP_006246848.1 | 9 - 10 | 1802 | 100 | ||
XM_006246789.2 | XP_006246851.1 | 8 - 9 | 1650 | 100 | ||
XM_008767857.1 | XP_008766079.1 | 9 - 10 | 1803 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
