Assay Details
Gene Symbol: Hsd17b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012851.2 | NP_036983.1 | 4 - 5 | 578 | 76 | |
GenBank mRNA | BC086365.1 | - | 4 - 5 | 563 | 76 | |
X78811.1 | - | 4 - 5 | 539 | 76 | ||
X97754.1 | - | 4 - 5 | 579 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
