Assay Details
Gene Symbol: Ccdc47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013974.1 | NP_001013996.1 | 11 - 12 | 1387 | 96 | |
GenBank mRNA | BC085824.1 | - | 11 - 12 | 1387 | 7127239 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
