Assay Details
Gene Symbol: Ccdc47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013974.1 | NP_001013996.1 | 1 - 2 | 160 | 133 | |
GenBank mRNA | BC085824.1 | - | 1 - 2 | 160 | 7127239 | 133 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC085824.1, NM_001013974.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
