Assay Details
Gene Symbol: Mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044282.1 | NP_001037747.1 | 2 - 3 | 342 | 113 | |
XM_006246126.2 | XP_006246188.1 | 2 - 3 | 492 | 113 | ||
GenBank mRNA | BC085899.1 | - | 2 - 3 | 342 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
